Thermo Scientific Chemicals

1,1'-Thiocarbonyldiimidazole, tech 90%, Thermo Scientific Chemicals

Catalog number: B22008.09
10 g, Each
Thermo Scientific Chemicals

1,1'-Thiocarbonyldiimidazole, tech 90%, Thermo Scientific Chemicals

Catalog number: B22008.09
10 g, Each
Quantity
Catalog number: B22008.09
also known as B22008-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6160-65-2
IUPAC Name
1-(1H-imidazole-1-carbothioyl)-1H-imidazole
Molecular Formula
C7H6N4S
InChI Key
RAFNCPHFRHZCPS-UHFFFAOYSA-N
SMILES
S=C(N1C=CN=C1)N1C=CN=C1
Appearance (Color)
Yellow
Assay (GC)
>88.0%
Purity (DSC)
>88.0% (non-U.S. specification)
Solution Test
(5% w/v in ethanol) : Clear
Form
Powder

Description

1,1'-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4'-dimethoxy-benzhydrol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4′-dimethoxy-benzhydrol.

Solubility
Soluble in terahydrofuran, toluene and dichloromethane.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text