Thermo Scientific Chemicals

Dimethyl pimelate, 98+%, Thermo Scientific Chemicals

Catalog number: B22026.06
5 g, Each
Thermo Scientific Chemicals

Dimethyl pimelate, 98+%, Thermo Scientific Chemicals

Catalog number: B22026.06
5 g, Each
Quantity
Catalog number: B22026.06
also known as B22026-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1732-08-7
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4290-1.4330 @ 20°C
Assay (GC)
≥98.5%

Description

Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl pimelate was used as the internal standard in the analysis of phthalates, sebacates, epoxidized soybean oil (ESBO) and polyadipates after transesterification to the ethyl esters.

Solubility
Soluble in ethanol, benzene, ether. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text