O-Methylisoureahemisulfate is used for the synthesis of fluorouracil antineoplastic and anthelmintic imidazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
O-Methylisoureahemisulfate is used for the synthesis of fluorouracil antineoplastic and anthelmintic imidazole.
Solubility
Soluble in organic solvents, water, methanol and ethanol
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Yang, W.; Kernstock, R.; Simmons, N.; Alak, A. ELISA microplate: a viable immunocapture platform over magnetic beads for immunoaffinity-LC-MS/MS quantitation of protein therapeutics?. Bioanalysis 2015, 7 (3), 307-318.
Cai, M.; Zhang, Z.; Su, X.; Dong, H.; Zhong, Z.; Zhuo, R. Guanidinated multi-arm star polyornithines with a polyethylenimine core for gene delivery. Polymer 2014, 55 (18), 4634-4640.