Thermo Scientific Chemicals

O-Methylisourea hemisulfate, 99%, Thermo Scientific Chemicals

Catalog number: B22036.30
250 g, Each
Thermo Scientific Chemicals

O-Methylisourea hemisulfate, 99%, Thermo Scientific Chemicals

Catalog number: B22036.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B22036.30
also known as B22036-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52328-05-9
IUPAC Name
methoxymethanimidamide
Molecular Formula
C2H6N2O
InChI Key
RMAHPRNLQIRHIJ-UHFFFAOYSA-N
SMILES
COC(N)=N
Melting Point (clear melt)
158.0-164.0?C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)
≤0.5%

Description

O-Methylisoureahemisulfate is used for the synthesis of fluorouracil antineoplastic and anthelmintic imidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Methylisoureahemisulfate is used for the synthesis of fluorouracil antineoplastic and anthelmintic imidazole.

Solubility
Soluble in organic solvents, water, methanol and ethanol

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text