Thermo Scientific Chemicals

tert-Butyl 2,2,2-trichloroacetimidate, 97%, Thermo Scientific Chemicals

Catalog number: B22039.14
25 g, Each
Thermo Scientific Chemicals

tert-Butyl 2,2,2-trichloroacetimidate, 97%, Thermo Scientific Chemicals

Catalog number: B22039.14
25 g, Each
Quantity
Catalog number: B22039.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98946-18-0
IUPAC Name
tert-butyl 2,2,2-trichloroethanimidate
Molecular Formula
C6H10Cl3NO
InChI Key
CQXDYHPBXDZWBA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=N)C(Cl)(Cl)Cl
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4535-1.4585 @ 20?C

Description

tert-Butyl 2,2,2-trichloroacetimidate is used to produce di-tert-butyl peroxide at temperature of -5°C. It may be used in the synthesis of N-α-Fmoc-phospho(1-nitrophenylethyl-2-cyanoethyl)-L-serine, caged building block. It may be used in the conversion of alcohols and carboxylic acids to their respective ethers and esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl 2,2,2-trichloroacetimidate is used to produce di-tert-butyl peroxide at temperature of -5°C. It may be used in the synthesis of N-α-Fmoc-phospho(1-nitrophenylethyl-2-cyanoethyl)-L-serine, caged building block. It may be used in the conversion of alcohols and carboxylic acids to their respective ethers and esters.

Solubility
Soluble in organic solvents, cyclo hexane.

Notes
Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text