Cobalt(II) hydroxide, 97%, Thermo Scientific Chemicals
Cobalt(II) hydroxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) hydroxide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number B22045.18
also known as B22045-18
Price (USD)/ Each
41.60
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
41.60
Add to cart
Cobalt(II) hydroxide, 97%, Thermo Scientific Chemicals
Catalog numberB22045.18
Price (USD)/ Each
41.60
-
Add to cart
Chemical Identifiers
CAS21041-93-0
IUPAC Nameλ²-cobalt(2+) dihydroxide
Molecular FormulaCoH2O2
InChI KeyASKVAEGIVYSGNY-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to dark pink
FormPowder
Assay (unspecified)>96.0 to <104.0% (Based on Co Elemental analysis)
Elemental Analysis(Cobalt, Co) : 60.87-65.94%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only