2-Methylheptanoic acid, 98%, Thermo Scientific Chemicals
2-Methylheptanoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylheptanoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B22050.14
also known as B22050-14
Price (USD)/ Each
81.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
81.00
Add to cart
2-Methylheptanoic acid, 98%, Thermo Scientific Chemicals
Catalog numberB22050.14
Price (USD)/ Each
81.00
-
Add to cart
Chemical Identifiers
CAS1188-02-9
IUPAC Name(2R)-2-methylheptanoate
Molecular FormulaC8H15O2
InChI KeyNKBWMBRPILTCRD-SSDOTTSWSA-M
SMILESCCCCC[C@@H](C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Refractive Index1.4235-1.4275 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only