1-Bromo-2-chloro-1,1,2-trifluoro-3-butene, 97%, Thermo Scientific Chemicals
1-Bromo-2-chloro-1,1,2-trifluoro-3-butene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-2-chloro-1,1,2-trifluoro-3-butene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22057.06
also known as B22057-06
Price (USD)/ Each
45.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
45.40
Add to cart
1-Bromo-2-chloro-1,1,2-trifluoro-3-butene, 97%, Thermo Scientific Chemicals
Catalog numberB22057.06
Price (USD)/ Each
45.40
-
Add to cart
Chemical Identifiers
CAS374-25-4
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4040-1.4090 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only