Thermo Scientific Chemicals

Sodium triacetoxyborohydride, 95%, Thermo Scientific Chemicals

Catalog number: B22060.06
5 g, Each
Thermo Scientific Chemicals

Sodium triacetoxyborohydride, 95%, Thermo Scientific Chemicals

Catalog number: B22060.06
5 g, Each
Quantity
Catalog number: B22060.06
also known as B22060-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56553-60-7
IUPAC Name
sodium bis(acetyloxy)boranuidyl acetate
Molecular Formula
C6H10BNaO6
InChI Key
HHYFEYBWNZJVFQ-UHFFFAOYSA-N
SMILES
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O
Form
Powder
Appearance (Color)
White
Assay from Supplier's CofA
≥94.0%

Description

Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text