Cobalt(II) phosphate, anhydrous, 98%, Thermo Scientific Chemicals
Cobalt(II) phosphate, anhydrous, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II) phosphate, anhydrous, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number B22066.22
also known as B22066-22
Price (USD)/ Each
178.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
178.00
Add to cart
Cobalt(II) phosphate, anhydrous, 98%, Thermo Scientific Chemicals
Catalog numberB22066.22
Price (USD)/ Each
178.00
-
Add to cart
Chemical Identifiers
CAS13455-36-2
IUPAC Nametris(λ²-cobalt(2+)) diphosphate
Molecular FormulaCo3O8P2
InChI KeyZBDSFTZNNQNSQM-UHFFFAOYSA-H
SMILES[Co++].[Co++].[Co++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Purple
FormPowder
Assay from Supplier's CofA≥97.5%
Trace MetalFe: <250ppm
Trace MetalNi: <2500ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only