Thermo Scientific Chemicals

Manganese(II) sulfate tetrahydrate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: B22081.22
100 g, Each
Thermo Scientific Chemicals

Manganese(II) sulfate tetrahydrate, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: B22081.22
100 g, Each
Quantity
Catalog number: B22081.22
also known as B22081-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10101-68-5
IUPAC Name
manganese(2+) sulfate
Molecular Formula
MnO4S
InChI Key
SQQMAOCOWKFBNP-UHFFFAOYSA-L
SMILES
[Mn++].[O-]S([O-])(=O)=O
Appearance (Color)
Pale pink to pink
Assay (unspecified)
Water: 29.5-34.9% (mole ratio 3.5 to 4.5:1)
Assay from Supplier's CofA
≥98.5%
Comment
May form fused solid on storage
Trace Metal
Total other metals ≤1.0%

Description

Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate tetrahydrate is an essential mineral used in capsule, tablet and liquid form, which provides essential nutrients like vitamin, protein and in similar nutritional substance. It undergoes electrolysis to give manganese dioxide. Upon oxidation, it gives chemical manganese dioxide (CMD) and finds application in dry- cell batteries.

Solubility
Soluble in water.

Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text