Thermo Scientific Chemicals

Sodium n-tridecyl sulfate, 99%, may cont. up to ca 5% water and residual solvent, Thermo Scientific Chemicals

Catalog number: B22089.14
25 g, Each
Thermo Scientific Chemicals

Sodium n-tridecyl sulfate, 99%, may cont. up to ca 5% water and residual solvent, Thermo Scientific Chemicals

Catalog number: B22089.14
25 g, Each
Quantity
Catalog number: B22089.14
also known as B22089-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3026-63-9
IUPAC Name
sodium tridecyl sulfate
Molecular Formula
C13H27NaO4S
InChI Key
HQCFDOOSGDZRII-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCCCOS([O-])(=O)=O
Appearance (Color)
White to pale cream
Form
Powder
Assay (ex Sulphated Ash)
>98.5% (dry wt. basis)
Water Content (Karl Fischer Titration)
≤5.0%
Loss on Drying
≤6.0%

Description

Sodium n-tridecyl sulfate acts as a foaming agent and a wetting agent. It is used as a cleaning agent and finds application in industrial and household cleaning products and personal care products. Further, it serves as an emulsifying agent in the transport of hydrocarbons. In addition to this, it acts as an anionic surfactant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-tridecyl sulfate acts as a foaming agent and a wetting agent. It is used as a cleaning agent and finds application in industrial and household cleaning products and personal care products. Further, it serves as an emulsifying agent in the transport of hydrocarbons. In addition to this, it acts as an anionic surfactant.

Solubility
Soluble in water.

Notes
Incompatible with bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text