Thermo Scientific Chemicals

Ficoll∣r 400, Thermo Scientific Chemicals

Catalog number: B22095.09
10 g, Each
Thermo Scientific Chemicals

Ficoll∣r 400, Thermo Scientific Chemicals

Catalog number: B22095.09
10 g, Each
Quantity
Catalog number: B22095.09
also known as B22095-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26873-85-8
IUPAC Name
2-(chloromethyl)oxirane; 2-{[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C15H27ClO12
InChI Key
IACFXVUNKCXYJM-UHFFFAOYNA-N
SMILES
ClCC1CO1.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(O)C1O
Form
Powder
Molecular Weight
300,000-500,000
Appearance (Color)
White
Assay from Supplier's CofA
≥98.0% (U.S. sourced material)
Loss on Drying
≤5.0% (non-U.S. sourced material); ≤7.0% (U.S. sourced material)

Description

Ficoll™ 400 is used for cell separation and organelle isolation. It is also used to separate blood to its components like erythrocytes and leukocytes. It acts as a mononuclear cell separator and used in culture media, nucleic acidhybridization, electrophoresis and in immunological studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text