4-Amino-2-chloropyridine, 98%, Thermo Scientific Chemicals
4-Amino-2-chloropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-2-chloropyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22098.06
also known as B22098-06
Price (USD)/ Each
67.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
67.50
Add to cart
4-Amino-2-chloropyridine, 98%, Thermo Scientific Chemicals
Catalog numberB22098.06
Price (USD)/ Each
67.50
-
Add to cart
Chemical Identifiers
CAS14432-12-3
IUPAC Name2-chloropyridin-4-amine
Molecular FormulaC5H5ClN2
InChI KeyBLBDTBCGPHPIJK-UHFFFAOYSA-N
SMILESNC1=CC=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow to cream to pale pink
FormCrystals or powder or crystalline powder
Melting Point (clear melt)88.0-94.5°C
Assay (GC)≥97.5%
2-Chloropyridin-4-amine (14432-12-3) is used as a medicine, pigment intermediates. It is an additive of plant growth regulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only