Thermo Scientific Chemicals

2-Fluoro-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22100.14
25 g, Each
Thermo Scientific Chemicals

2-Fluoro-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22100.14
25 g, Each
Quantity
Catalog number: B22100.14
also known as B22100-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7304-32-7
Melting Point (clear melt)
137.0-143.0?C
Appearance (Color)
Cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%

Description

2-Fluoro-5-nitrobenzoic acid may be used in the synthesis of dibenz[b,f]oxazepin-11(10H)-ones, via the nucleophilic aromatic substitution (SNAr) of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH of various 2-aminophenols on solid support, synthesis of substituted dibenzazocines on solid support, via SNAr of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH function of the immobilized polysubstituted phenols, in the synthesis of oxazepines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-nitrobenzoic acid may be used in the synthesis of dibenz[b,f]oxazepin-11(10H)-ones, via the nucleophilic aromatic substitution (SNAr) of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH of various 2-aminophenols on solid support, synthesis of substituted dibenzazocines on solid support, via SNAr of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH function of the immobilized polysubstituted phenols, in the synthesis of oxazepines.

Solubility
Soluble in chloroform, ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text