Tricarballylic acid, 98%, Thermo Scientific Chemicals
Tricarballylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tricarballylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B22103.14
also known as B22103-14
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
Add to cart
Tricarballylic acid, 98%, Thermo Scientific Chemicals
Catalog numberB22103.14
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS99-14-9
IUPAC Namepropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O6
InChI KeyKQTIIICEAUMSDG-UHFFFAOYSA-N
SMILESOC(=O)CC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)154-160?C
It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

Solubility
Soluble in water (500 mg/ml at 18°C), methanol, and ether (slightly).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Craig D Boyle; Yoshito Kishi. Absolute configuration at the tricarballylic acid moieties of fumonisin B2. Tetrahedron Letters. 1995, 36 (26), 4579-4582.
  2. Frank C. Magne; Robert R. Mod. Plasticizers from Aconitic and Tricarballylic Acids. Ind. Eng. Chem. 1953, 45 (7), 1546-1547.