Dowex™ 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dowex|r 50WX8 50-100 (H) is used in deionizing supernatant fraction samples for paper chromatography and as a catalyst. It is involved in the separation medium for thin layer chromatography, paper chromatography and low pressure liquid chromatography.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
Strongly acidic cation-exchange resin, 8% cross-linked, H+ form, 50-100 mesh (dry).
Kang, Y. G.; Park, C. Y.; Shin, H.; Singh, R.; Arora, G.; Yu, C. M.; Lee, I. Y. Synthesis and anti-tubercular activity of 2-nitroimidazooxazines with modification at the C-7 position as PA-824 analogs. Bioorg. Med. Chem. Lett. 2015, 25 (17), 3650-3653.
Chung, K. H.; Kim, H.; Lim, J. M.; Ji, Y. Y.; Choi, G. S.; Kang, M. J. Rapid determination of radiostrontium in milk using automated radionuclides separator and liquid scintillation counter. J. Radioanal. Nucl. Chem. 2015, 304 (1), 293-300.