Thermo Scientific Chemicals

Trifluoromethanesulfonamide, 96%, Thermo Scientific Chemicals

Catalog number: B22112.03
1 g, Each
Thermo Scientific Chemicals

Trifluoromethanesulfonamide, 96%, Thermo Scientific Chemicals

Catalog number: B22112.03
1 g, Each
Quantity
Catalog number: B22112.03
also known as B22112-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
421-85-2
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
115.0-124.0?C
Appearance (Color)
White to pale cream

Description

Trifluoromethanesulfonamide is used as an intermediate in synthetic chemistry. It is used in the preparation of specific inhibitors of mammalian secreted phospholipases A2. It is also involved in the synthesis of ecicosanoids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoromethanesulfonamide is used as an intermediate in synthetic chemistry. It is used in the preparation of specific inhibitors of mammalian secreted phospholipases A2. It is also involved in the synthesis of ecicosanoids.

Solubility
Soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text