4-Hydroxy-4'-iodobiphenyl, 98+%, Thermo Scientific Chemicals
4-Hydroxy-4'-iodobiphenyl, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-4'-iodobiphenyl, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22115.06
also known as B22115-06
Price (USD)/ Each
68.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
68.90
Add to cart
4-Hydroxy-4'-iodobiphenyl, 98+%, Thermo Scientific Chemicals
Catalog numberB22115.06
Price (USD)/ Each
68.90
-
Add to cart
Chemical Identifiers
CAS29558-78-9
IUPAC Name4'-iodo-[1,1'-biphenyl]-4-ol
Molecular FormulaC12H9IO
InChI KeyWJXIAMCDWSUSEI-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point196-203°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only