Thermo Scientific Chemicals

Methyl 3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: B22129.06
5 g, Each
Thermo Scientific Chemicals

Methyl 3-hydroxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: B22129.06
5 g, Each
Quantity
Catalog number: B22129.06
also known as B22129-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5118-06-9
IUPAC Name
(2Z)-2-[hydroxy(methoxy)methylidene]-2,3-dihydrothiophen-3-one
Molecular Formula
C6H6O3S
InChI Key
FYOPNADYSPZUAT-WAYWQWQTSA-N
SMILES
COC(O)=C1/SC=CC1=O
Assay (GC)
≥96.0%
Appearance (Color)
White to pale yellow or pale cream to pale brown
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
41.0-47.0?C

Description

Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-hydroxythiophene-2-carboxylate is used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text