Thermo Scientific Chemicals

N-Phenylethanolamine, 98%, Thermo Scientific Chemicals

Catalog number: B22131.36
500 g, Each
Thermo Scientific Chemicals

N-Phenylethanolamine, 98%, Thermo Scientific Chemicals

Catalog number: B22131.36
500 g, Each
Quantity
Catalog number: B22131.36
also known as B22131-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-98-5
IUPAC Name
2-(phenylamino)ethan-1-ol
Molecular Formula
C8H11NO
InChI Key
MWGATWIBSKHFMR-UHFFFAOYSA-N
SMILES
OCCNC1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow to brown
Assay (GC)
≥97.5%
Refractive Index
1.5780-1.5820 @ 20?C
Form
Liquid

Description

N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5'-diphospho-glucuronosyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5′-diphospho-glucuronosyltransferase.

Solubility
Slightly miscible with water.

Notes
Incompatible with acids, acid anhydrides, acid chlorides and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text