Thermo Scientific Chemicals

alpha-D-Glucose pentaacetate, 99%, Thermo Scientific Chemicals

Catalog number: B22137.18
50 g, Each
Thermo Scientific Chemicals

alpha-D-Glucose pentaacetate, 99%, Thermo Scientific Chemicals

Catalog number: B22137.18
50 g, Each
Quantity
Catalog number: B22137.18
also known as B22137-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
604-68-2
IUPAC Name
[3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
Molecular Formula
C16H22O11
InChI Key
LPTITAGPBXDDGR-UHFFFAOYNA-N
SMILES
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Appearance (Color)
White
Form
Powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
108.0-115.0?C
Optical Rotation
102.0 ± 2.5? (C=5 in chloroform)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text