Thermo Scientific Chemicals

alpha-Toluenesulfonyl fluoride, 99%, Thermo Scientific Chemicals

Catalog number: B22146.03
1 g, Each
Thermo Scientific Chemicals

alpha-Toluenesulfonyl fluoride, 99%, Thermo Scientific Chemicals

Catalog number: B22146.03
1 g, Each
Quantity
Catalog number: B22146.03
also known as B22146-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
329-98-6
IUPAC Name
phenylmethanesulfonyl fluoride
Molecular Formula
C7H7FO2S
InChI Key
YBYRMVIVWMBXKQ-UHFFFAOYSA-N
SMILES
FS(=O)(=O)CC1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point
91-96?C

Description

α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.

Solubility
Soluble in ether, benzene, anhydrous ethanol, methanol, 2- propane. Hydrolyses in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, amines, bases, water/ moisture. Store at 4°C
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text