Thermo Scientific Chemicals

4'-Methoxychalcone, 97%, Thermo Scientific Chemicals

Catalog number: B22158.06
5 g, Each
Thermo Scientific Chemicals

4'-Methoxychalcone, 97%, Thermo Scientific Chemicals

Catalog number: B22158.06
5 g, Each
Quantity
Catalog number: B22158.06
also known as B22158-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
959-23-9
IUPAC Name
(2E)-1-(4-methoxyphenyl)-3-phenylprop-2-en-1-one
Molecular Formula
C16H14O2
InChI Key
KJHHAPASNNVTSN-KPKJPENVSA-N
SMILES
COC1=CC=C(C=C1)C(=O)C=CC1=CC=CC=C1
Appearance (Color)
White to pale cream to pale yellow
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
102.0-111.0?C

Description

4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.

Solubility
Soluble in dichloromethane and methanol. Insoluble in water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text