Ethyl 4-bromophenylacetate, 98%, Thermo Scientific Chemicals
Ethyl 4-bromophenylacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-bromophenylacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B22165.22
also known as B22165-22
Price (USD)/ Each
459.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
459.00
Add to cart
Ethyl 4-bromophenylacetate, 98%, Thermo Scientific Chemicals
Catalog numberB22165.22
Price (USD)/ Each
459.00
-
Add to cart
Chemical Identifiers
CAS14062-25-0
IUPAC Nameethyl 2-(4-bromophenyl)acetate
Molecular FormulaC10H11BrO2
InChI KeyZFDCWHPNBWPPHG-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
FormFused solid
Melting Point (clear melt)28.0-34.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only