4-tert-Butylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
4-tert-Butylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-tert-Butylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number B22174.18
also known as B22174-18
Price (USD)/ Each
139.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
139.00
Add to cart
4-tert-Butylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberB22174.18
Price (USD)/ Each
139.00
-
Add to cart
Chemical Identifiers
CAS15084-51-2
IUPAC Name4-tert-butylbenzene-1-sulfonyl chloride
Molecular FormulaC10H13ClO2S
InChI KeyYEZADZMMVHWFIY-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)78.0-85.0°C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only