3-Amino-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals
3-Amino-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B22176.22
also known as B22176-22
Price (USD)/ Each
993.65
Online exclusive
1104.00 
Save 110.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
993.65
Online exclusive
1104.00 
Save 110.35 (10%)
Add to cart
3-Amino-3-(2-nitrophenyl)propionic acid, 98%, Thermo Scientific Chemicals
Catalog numberB22176.22
Price (USD)/ Each
993.65
Online exclusive
1104.00 
Save 110.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5678-48-8
IUPAC Name(3R)-3-azaniumyl-3-(2-nitrophenyl)propanoate
Molecular FormulaC9H10N2O4
InChI KeyXXBOYULKNZTOMN-SSDOTTSWSA-N
SMILES[NH3+][C@H](CC([O-])=O)C1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-3-(2-nitrophenyl)propionic acid is used in chemical research.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 0-5°C. Incompatible with strong oxidizing agents, acids, bases.
RUO – Research Use Only
Bradley B. Brown; David S. Wagner; H. Mario Geysen. A single-bead decode strategy using electrospray ionization mass spectrometry and a new photolabile linker: 3-Amino-3-(2-nitrophenyl)propionic acid. Molecular Diversity. 1995, 1 (1), 4-12.