1-Methylpiperidine-2-methanol, 97%, Thermo Scientific Chemicals
1-Methylpiperidine-2-methanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methylpiperidine-2-methanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B22187.22
also known as B22187-22
Price (USD)/ Each
201.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
201.00
Add to cart
1-Methylpiperidine-2-methanol, 97%, Thermo Scientific Chemicals
Catalog numberB22187.22
Price (USD)/ Each
201.00
-
Add to cart
Chemical Identifiers
CAS20845-34-5
IUPAC Name(1-methylpiperidin-2-yl)methanol
Molecular FormulaC7H15NO
InChI KeyHXXJMMLIEYAFOZ-UHFFFAOYNA-N
SMILESCN1CCCCC1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow to brown
Refractive Index1.4805-1.4855 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only