1,4-Bis(difluoromethyl)benzene, 98%, Thermo Scientific Chemicals
1,4-Bis(difluoromethyl)benzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Bis(difluoromethyl)benzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22193.03
also known as B22193-03
Price (USD)/ Each
41.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
41.60
Add to cart
1,4-Bis(difluoromethyl)benzene, 98%, Thermo Scientific Chemicals
Catalog numberB22193.03
Price (USD)/ Each
41.60
-
Add to cart
Chemical Identifiers
CAS369-54-0
IUPAC Name1,4-bis(difluoromethyl)benzene
Molecular FormulaC8H6F4
InChI KeyVWKMZVCSRVFUGW-UHFFFAOYSA-N
SMILESFC(F)C1=CC=C(C=C1)C(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4380-1.4420 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only