Thermo Scientific Chemicals

Methyl difluoroacetate, 99%, Thermo Scientific Chemicals

Catalog number: B22197.06
5 g, Each
Thermo Scientific Chemicals

Methyl difluoroacetate, 99%, Thermo Scientific Chemicals

Catalog number: B22197.06
5 g, Each
Quantity
Catalog number: B22197.06
also known as B22197-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
433-53-4
IUPAC Name
methyl 2,2-difluoroacetate
Molecular Formula
C3H4F2O2
InChI Key
CSSYKHYGURSRAZ-UHFFFAOYSA-N
SMILES
COC(=O)C(F)F
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.3335-1.3375 @ 20?C

Description

A methyl difluoroacetate (MFA)-based LiPF6 solution was applied as an electrolyte to improve the thermal stability of Li-ion batteries. Methyl difluoroacetate is used in the inhibition of corrosion of aluminum cathode current collector for lithium ion cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A methyl difluoroacetate (MFA)-based LiPF6 solution was applied as an electrolyte to improve the thermal stability of Li-ion batteries. Methyl difluoroacetate is used in the inhibition of corrosion of aluminum cathode current collector for lithium ion cells.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text