Thermo Scientific Chemicals

2,2-Difluoroethanol, 97%, Thermo Scientific Chemicals

Catalog number: B22201.03
1 g, Each
Thermo Scientific Chemicals

2,2-Difluoroethanol, 97%, Thermo Scientific Chemicals

Catalog number: B22201.03
1 g, Each
Quantity
Catalog number: B22201.03
also known as B22201-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
359-13-7
IUPAC Name
2,2-difluoroethan-1-ol
Molecular Formula
C2H4F2O
InChI Key
VOGSDFLJZPNWHY-UHFFFAOYSA-N
SMILES
OCC(F)F
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.3310-1.3360 @ 20?C

Description

2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

Solubility
soluble in chloroform, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text