2,2-Difluoroethanol, 97%, Thermo Scientific Chemicals
2,2-Difluoroethanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2-Difluoroethanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B22201.06
also known as B22201-06
Price (USD)/ Each
217.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
217.00
Add to cart
2,2-Difluoroethanol, 97%, Thermo Scientific Chemicals
Catalog numberB22201.06
Price (USD)/ Each
217.00
-
Add to cart
Chemical Identifiers
CAS359-13-7
IUPAC Name2,2-difluoroethan-1-ol
Molecular FormulaC2H4F2O
InChI KeyVOGSDFLJZPNWHY-UHFFFAOYSA-N
SMILESOCC(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.3310-1.3360 @ 20?C
2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

Solubility
soluble in chloroform, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, acids.
RUO – Research Use Only

General References:

  1. Matti Perttilä. Vibrational spectra and normal coordinate analysis of 2,2-difluoroethanol and 2,2-dichloroethanol. Spectrochimica Acta Part A: Molecular Spectroscopy. 1979, 35 (1), 37-44.
  2. R. Malhotra; L. A. Woolf. Thermodynamic properties of liquid 2,2-difluoroethanol. International Journal of Thermophysics. 1995, 16 (4), 901-908.
  3. Precursor of a monofluoro acyl anion equivalent; compare 2,2,2-Trifluoroethanol, A10788: Synlett, 503 (1994).