Thermo Scientific Chemicals

Diethoxydiphenylsilane, 98%, Thermo Scientific Chemicals

Catalog number: B22204.30
250 g, Each
Thermo Scientific Chemicals

Diethoxydiphenylsilane, 98%, Thermo Scientific Chemicals

Catalog number: B22204.30
250 g, Each
Quantity
Catalog number: B22204.30
also known as B22204-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2553-19-7
IUPAC Name
diethoxydiphenylsilane
Molecular Formula
C16H20O2Si
InChI Key
ZZNQQQWFKKTOSD-UHFFFAOYSA-N
SMILES
CCO[Si](OCC)(C1=CC=CC=C1)C1=CC=CC=C1
Refractive Index
1.5240-1.5290 @ 20?C
Appearance (Color)
Clear colorless to dark yellow
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Form
Liquid

Description

Reagent for preparation of diphenylsilylene derivatives of diols by exchange.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text