4-Methylthio-2-butanone, 98%, Thermo Scientific Chemicals
4-Methylthio-2-butanone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylthio-2-butanone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22206.06
also known as B22206-06
Price (USD)/ Each
58.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
58.70
Add to cart
4-Methylthio-2-butanone, 98%, Thermo Scientific Chemicals
Catalog numberB22206.06
Price (USD)/ Each
58.70
-
Add to cart
Chemical Identifiers
CAS34047-39-7
IUPAC Name4-(methylsulfanyl)butan-2-one
Molecular FormulaC5H10OS
InChI KeyDRGHCRKOWMAZAO-UHFFFAOYSA-N
SMILESCSCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4725-1.4765 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only