Thermo Scientific Chemicals

Sodium 1-propanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: B22210.14
25 g, Each
Thermo Scientific Chemicals

Sodium 1-propanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: B22210.14
25 g, Each
Quantity
Catalog number: B22210.14
also known as B22210-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14533-63-2
IUPAC Name
sodium propane-1-sulfonate
Molecular Formula
C3H7NaO3S
InChI Key
NPAWNPCNZAPTKA-UHFFFAOYSA-M
SMILES
[Na+].CCCS([O-])(=O)=O
Appearance (Color)
White
Form
Powder
Assay (unspecified)
≥98.5% (dry wt. basis)
Water Content (Karl Fischer Titration)
<1.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text