Thermo Scientific Chemicals

4-Ethyloctanoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22220.88
2.5 g, Each
Thermo Scientific Chemicals

4-Ethyloctanoic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22220.88
2.5 g, Each
Quantity
Catalog number: B22220.88
also known as B22220-88
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16493-80-4
IUPAC Name
(4R)-4-ethyloctanoate
Molecular Formula
C10H19O2
InChI Key
PWKJMPFEQOHBAC-SECBINFHSA-M
SMILES
CCCC[C@@H](CC)CCC([O-])=O
Appearance (Color)
Clear colorless
Refractive Index
1.4360-1.4410 @ 20?C
Form
Liquid
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Identification (FTIR)
Conforms

Description

Five sulfur-containing flavor compounds were synthesized by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. 4-ethyloctanoic acid is identified from flue-cured Virginia tobacco. Mainly used as daily flavor,food flavor,industrial flavor, nutrient, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Five sulfur-containing flavor compounds were synthesized by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. 4-ethyloctanoic acid is identified from flue-cured Virginia tobacco. Mainly used as daily flavor,food flavor,industrial flavor, nutrient, stabilizers, surfactants and emulsifiers.

Solubility
Slightly soluble in water(0.13 mg/mL). Soluble in hexane

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text