Thermo Scientific Chemicals

Acid Fuchsin sodium salt

Catalog number: B22222.22
100 g, Each
Thermo Scientific Chemicals

Acid Fuchsin sodium salt

Catalog number: B22222.22
100 g, Each
Quantity
Catalog number: B22222.22
also known as B22222-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3244-88-0
IUPAC Name
disodium 2-amino-5-[(4-amino-3-sulfophenyl)[(1Z)-4-imino-3-methyl-5-sulfonatocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Molecular Formula
C20H17N3Na2O9S3
InChI Key
WZRZTHMJPHPAMU-UHFFFAOYSA-L
SMILES
[Na+].[Na+].CC1=C\C(C=C(C1=N)S([O-])(=O)=O)=C(\C1=CC=C(N)C(=C1)S(O)(=O)=O)C1=CC=C(N)C(=C1)S([O-])(=O)=O
lambda-max (UV-Vis)
540 - 550nm (water)
Appearance (Color)
Dark green to dark brown
Form
Crystals or powder or crystalline powder

Description

Acid Fuchsin sodium salt is used as a pH indicator. It is one of the dyes used in Masson’s Trichome Stain where it is used to distinguish muscle from collagen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acid Fuchsin sodium salt is used as a pH indicator. It is one of the dyes used in Masson’s Trichome Stain where it is used to distinguish muscle from collagen.

Solubility
Soluble in water (1 mg/ml).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text