Thermo Scientific Chemicals

2,3-Dimethylbutane, 99%, Thermo Scientific Chemicals

Catalog number: B22223.22
100 g, Each
Thermo Scientific Chemicals

2,3-Dimethylbutane, 99%, Thermo Scientific Chemicals

Catalog number: B22223.22
100 g, Each
Quantity
Catalog number: B22223.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-29-8
IUPAC Name
2,3-dimethylbutane
Molecular Formula
C6H14
InChI Key
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
SMILES
CC(C)C(C)C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.3735-1.3765 @ 20?C
Form
Liquid

Description

2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethylbutane is a branched saturated hydrocarbon for proteomics research. It is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Soluble in water (1mg/ml @22°C), DMSO, ethanol, acetone.

Notes
Incompatible with strong oxidizing agents, oxygen, ammonia.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text