Thermo Scientific Chemicals

Sodium nicotinate, 98%, Thermo Scientific Chemicals

Catalog number: B22224.36
500 g, Each
Thermo Scientific Chemicals

Sodium nicotinate, 98%, Thermo Scientific Chemicals

Catalog number: B22224.36
500 g, Each
Quantity
Catalog number: B22224.36
also known as B22224-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54-86-4
IUPAC Name
sodium pyridine-3-carboxylate
Molecular Formula
C6H4NNaO2
InChI Key
KFLRWGSAMLBHBV-UHFFFAOYSA-M
SMILES
[Na+].[O-]C(=O)C1=CC=CN=C1
Melting Point
>300?C
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text