Thermo Scientific Chemicals

4-Bromo-3-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: B22227.09
10 g, Each
Thermo Scientific Chemicals

4-Bromo-3-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: B22227.09
10 g, Each
Quantity
Catalog number: B22227.09
also known as B22227-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
656-65-5
IUPAC Name
4-bromo-3-fluoroaniline
Molecular Formula
C6H5BrFN
InChI Key
YTMVYYAKOPIJCZ-UHFFFAOYSA-N
SMILES
NC1=CC=C(Br)C(F)=C1
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
65.0-71.0?C

Description

4-Bromo-3-fluoroaniline is an intermediate used for the synthetic preparation of Tradizolid and various pharmaceuticals such as the synthesis of several lateral difluoro-substituted 4,4-dialkyl- and 4,4-alkoxyalkylterphenyls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-3-fluoroaniline is an intermediate used for the synthetic preparation of Tradizolid and various pharmaceuticals such as the synthesis of several lateral difluoro-substituted 4,4″-dialkyl- and 4,4″-alkoxyalkylterp
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text