Thermo Scientific Chemicals

3-Chloro-2,6-dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: B22232.30
250 g, Each
Thermo Scientific Chemicals

3-Chloro-2,6-dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: B22232.30
250 g, Each
Quantity
Catalog number: B22232.30
also known as B22232-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26829-77-6
IUPAC Name
3-chloro-2,6-dimethylaniline
Molecular Formula
C8H10ClN
InChI Key
FGMXFOTYCHZCLA-UHFFFAOYSA-N
SMILES
CC1=CC=C(Cl)C(C)=C1N
Appearance (Color)
White to dark brown to pale brown or pink
Form
Fused solid or clear liquid as melt
Assay (GC)
≥98.5%
Melting Point (clear melt)
28.0-35.0?C

Description

3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-cyano- 2,4-dimethylchlorobenzene was obtained from 3-Chloro-2,6-dimethylaniline which upon hydrolysis gives acid.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text