Thermo Scientific Chemicals

BAPTA tetrasodium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog number: B22258.14
25 g, Each
Thermo Scientific Chemicals

BAPTA tetrasodium salt hydrate, 98%, Thermo Scientific Chemicals

Catalog number: B22258.14
25 g, Each
Quantity
Catalog number: B22258.14
also known as B22258-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
336624-09-0
IUPAC Name
tetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Molecular Formula
C22H20N2Na4O10
InChI Key
ZWSMLJACYSGFKD-UHFFFAOYSA-J
SMILES
[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=C(OCCOC2=C(C=CC=C2)N(CC([O-])=O)CC([O-])=O)C=CC=C1
Elemental Analysis
Magnesium ≤50ppm
Water Content (Karl Fischer Titration)
5.4-6.6% (dihydrate 6.0% water by weight)
Proton NMR
Conforms to structure
Appearance (Color)
White to pale cream
Complexometric Titration
≥97.5 to ≤102.5% (dry wt. basis)

Description

BAPTA tetrasodium salt hydrateis a high quality and sensitive compound for calcium signaling studies(investigation of signal transduction and apoptotic cascades, and neuroscience research). BAPTA, Tetrasodium Salt is useful for spectrophotometric monitoring of extracellular Ca2+ levels, with very little sensitivity to pH. Has also been used for the measurement and control of Ca2+ levels in eggs, chromaffin cells, muscle cells, retinal cells, and pancreatic cells. BAPTA-Na4 is a fluorescent compound utilized to determine free Ca2+ by performing fluorescence emission scans at an excitation wavelength of 299 nm and measuring the emission wavelength at 360 nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BAPTA tetrasodium salt hydrateis a high quality and sensitive compound for calcium signaling studies(investigation of signal transduction and apoptotic cascades, and neuroscience research). BAPTA, Tetrasodium Salt is useful for spectrophotometric monitoring of extracellular Ca2+ levels, with very little sensitivity to pH. Has also been used for the measurement and control of Ca2+ levels in eggs, chromaffin cells, muscle cells, retinal cells, and pancreatic cells. BAPTA-Na4 is a fluorescent compound utilized to determine free Ca2+ by performing fluorescence emission scans at an excitation wavelength of 299 nm and measuring the emission wavelength at 360 nm.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text