Ethyl 2-chloro-4,4,4-trifluoroacetoacetate, 94%, Thermo Scientific Chemicals
Ethyl 2-chloro-4,4,4-trifluoroacetoacetate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-chloro-4,4,4-trifluoroacetoacetate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22270.06
also known as B22270-06
Price (USD)/ Each
65.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
65.70
Add to cart
Ethyl 2-chloro-4,4,4-trifluoroacetoacetate, 94%, Thermo Scientific Chemicals
Catalog numberB22270.06
Price (USD)/ Each
65.70
-
Add to cart
Chemical Identifiers
CAS363-58-6
IUPAC Nameethyl (2S)-2-chloro-4,4,4-trifluoro-3-oxobutanoate
Molecular FormulaC6H6ClF3O3
InChI KeyYVWUNJVPOCYLIM-VKHMYHEASA-N
SMILESCCOC(=O)[C@@H](Cl)C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Appearance (Color)Clear, colourless to yellow
Assay (GC)> 92.5% (sum of keto + enol forms)
Refractive Index1.3865 - 1.3935 @20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only