Thermo Scientific Chemicals

Diethyl 4-aminobenzylphosphonate, 99%, Thermo Scientific Chemicals

Catalog number: B22279.06
5 g, Each
Thermo Scientific Chemicals

Diethyl 4-aminobenzylphosphonate, 99%, Thermo Scientific Chemicals

Catalog number: B22279.06
5 g, Each
Quantity
Catalog number: B22279.06
also known as B22279-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20074-79-7
IUPAC Name
diethyl [(4-aminophenyl)methyl]phosphonate
Molecular Formula
C11H18NO3P
InChI Key
ZVAYUUUQOCPZCZ-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC1=CC=C(N)C=C1)OCC
Assay (GC)
≥98.5%
Melting Point (clear melt)
88.0-95.0?C
Appearance (Color)
White to cream to yellow to pale brown
Form
Crystals or powder or crystalline powder

Description

Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 4-aminobenzylphosphonate is used as a reagent in the preparation of [4-(tetradecanoylamino)benzyl]phosphonic acid. It is also used in the production of diethyl p-benzoylaminobenzylphosphonate.

Solubility
Soluble in methylene chloride, acetone and benzene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text