Thermo Scientific Chemicals

3,5-Dimethoxyphenol, 98%, Thermo Scientific Chemicals

Catalog number: B22283.18
50 g, Each
Thermo Scientific Chemicals

3,5-Dimethoxyphenol, 98%, Thermo Scientific Chemicals

Catalog number: B22283.18
50 g, Each
Quantity
Catalog number: B22283.18
also known as B22283-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
500-99-2
Appearance (Color)
White to pale brown or yellow
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥97.5%
Melting Point
40.0-46.0?C

Description

3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-dimethoxyphenol is a marker for poisoning from Taxus baccata.

Solubility
Soluble in methanol, ethanol, benzene, water (slightly), and chloroform.

Notes
Air and light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text