Thermo Scientific Chemicals

Iodonitrotetrazolium violet, 95%

Catalog number: B22301.03
1 g, Each
Thermo Scientific Chemicals

Iodonitrotetrazolium violet, 95%

Catalog number: B22301.03
1 g, Each
Quantity
Catalog number: B22301.03
also known as B22301-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
146-68-9
IUPAC Name
3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Formula
C19H13ClIN5O2
InChI Key
JORABGDXCIBAFL-UHFFFAOYSA-M
SMILES
[Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1
Form
Powder
lambda-max (UV-Vis)
245-249nm
Appearance (Color)
Cream to yellow
Assay (Titration ex Chloride)
≥94.0 to ≤106.0% (dry weight basis)
Loss on Drying
≤6.0% (105?C/constant weight)

Description

Used for dehydrogenase determination

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text