Thermo Scientific Chemicals

2,2'-Thiodiethanethiol, tech. 85%, Thermo Scientific Chemicals

Catalog number: B22304.30
250 g, Each
Thermo Scientific Chemicals

2,2'-Thiodiethanethiol, tech. 85%, Thermo Scientific Chemicals

Catalog number: B22304.30
250 g, Each
Quantity
Catalog number: B22304.30
also known as B22304-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3570-55-6
IUPAC Name
2-[(2-sulfanylethyl)sulfanyl]ethane-1-thiol
Molecular Formula
C4H10S3
InChI Key
KSJBMDCFYZKAFH-UHFFFAOYSA-N
SMILES
SCCSCCS
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥80.0%
Refractive Index
1.5555-1.6355 @ 20°C

Description

2,2?-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2'-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2′-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text