Dibromomalonamide is an antimicrobial agent used in sugar mills, a compound for lipophilic fluid systems for fabric care. It is also used to produce 5-thioxo-[1,3]dithiolo[4,5-d][1,3]dithiole-2,2-dicarboxylic acid diamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibromomalonamide is an antimicrobial agent used in sugar mills, a compound for lipophilic fluid systems for fabric care. It is also used to produce 5-thioxo-[1,3]dithiolo[4,5-d][1,3]dithiole-2,2-dicarboxylic acid diamide.
Solubility
Soluble in water.
Notes
Moisture & light sensitive. Store away from oxidizing agent.
RUO – Research Use Only
Teruaki Mukaiyama; Tsujiaki Hata; Koa Tasaka. A Selective Phosphorylation by Means of Dibromomalonamide and Trialkyl Phosphites. J. Org. Chem. 1963, 28 (2), 481-483.
Hans Zimmer; Gurdial Singh. Synthesis of Some Triphenylphosphinalkylimines and Mono- and Dialkylaminotriphenylphosphonium Halides. J. Org. Chem. 1963, 28 (2), 483-486.
The combination with triethyl phosphite is a useful reagent for the phosphorylation of alcohols: J. Org. Chem., 28, 481 (1963):