Thermo Scientific Chemicals

4-n-Butylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: B22316.14
25 g, Each
Thermo Scientific Chemicals

4-n-Butylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog number: B22316.14
25 g, Each
Quantity
Catalog number: B22316.14
also known as B22316-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57802-79-6
IUPAC Name
(4-butylphenyl)methanaminium
Molecular Formula
C11H18N
InChI Key
IBVGSPOHLFKLHM-UHFFFAOYSA-O
SMILES
CCCCC1=CC=C(C[NH3+])C=C1
Comment
Placeholder record added to rectify the version number

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text