Thermo Scientific Chemicals

1-(p-Toluenesulfonyl)indole, 95%, Thermo Scientific Chemicals

Catalog number: B22320.03
1 g, Each
Thermo Scientific Chemicals

1-(p-Toluenesulfonyl)indole, 95%, Thermo Scientific Chemicals

Catalog number: B22320.03
1 g, Each
Quantity
Catalog number: B22320.03
also known as B22320-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31271-90-6
IUPAC Name
1-(4-methylbenzenesulfonyl)-1H-indole
Molecular Formula
C15H13NO2S
InChI Key
JNRRPYFLDADLJW-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12
Appearance (Color)
White to pale cream
Melting Point (clear melt)
79.0-91.0?C
Assay (GC)
≥94.0%
Form
Powder

Description

1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text